Assay Details
| Gene Symbol: elavl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244600.1 | NP_001231529.1 | 3 - 4 | 861 | 61 | |
| NM_001244601.1 | NP_001231530.1 | 3 - 4 | 735 | 61 | ||
| NM_131453.3 | NP_571528.2 | 3 - 4 | 496 | 61 | ||
| XM_005166786.4 | XP_005166843.1 | 2 - 3 | 773 | 61 | ||
| XM_017357272.2 | XP_017212761.1 | 2 - 3 | 612 | 61 | ||
| XM_021478178.1 | XP_021333853.1 | 3 - 4 | 859 | 61 | ||
| XM_021478179.1 | XP_021333854.1 | 3 - 4 | 859 | 61 | ||
| XM_021478180.1 | XP_021333855.1 | 3 - 4 | 859 | 61 | ||
| GenBank mRNA | BC065965.1 | - | 3 - 4 | 496 | 61 | |
| U17602.1 | - | 2 - 3 | 342 | 61 | ||
More Information
Set Membership: |
|

elavl4
Genomic Map