Assay Details
| Gene Symbol: trpc4apb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001310093.1 | NP_001297022.1 | 15 - 16 | 1764 | 87 | |
| XM_017353602.2 | XP_017209091.1 | 15 - 16 | 1773 | 87 | ||
| GenBank mRNA | AF130460.1 | - | 15 - 16 | 1784 | 87 | |
| BC162514.1 | - | 15 - 16 | 1764 | 87 | ||
More Information
Set Membership: |
|

trpc4apb
Genomic Map