Assay Details
| Gene Symbol: trpc4apb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017353602.2 | XP_017209091.1 | 17 - 18 | 2004 | 106 | |
| GenBank mRNA | AF130460.1 | - | 17 - 18 | 104 | ||
More Information
Set Membership: |
|

trpc4apb
Genomic Map