Assay Details
Gene Symbol: trpc4apb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001310093.1 | NP_001297022.1 | 6 - 7 | 637 | 127 | |
XM_017353602.2 | XP_017209091.1 | 6 - 7 | 646 | 127 | ||
GenBank mRNA | AF130460.1 | - | 6 - 7 | 657 | 127 | |
BC162514.1 | - | 6 - 7 | 637 | 127 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
