Assay Details
Gene Symbol: pax2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131640.1 | NP_571715.1 | 3 - 4 | 852 | 87 | |
GenBank mRNA | AF072547.1 | - | 3 - 4 | 852 | 87 | |
BC162214.1 | - | 3 - 4 | 553 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
