Assay Details
| Gene Symbol: pax2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131640.1 | NP_571715.1 | 8 - 9 | 1446 | 84 | |
| GenBank mRNA | AF072547.1 | - | 8 - 9 | 1446 | 84 | |
| BC162214.1 | - | 8 - 9 | 1147 | 84 | ||
More Information
Set Membership: |
|

pax2b
Genomic Map