Assay Details
| Gene Symbol: mapk8b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131721.2 | NP_571796.1 | 2 - 3 | 297 | 54 | |
| XM_017358485.2 | XP_017213974.1 | 2 - 3 | 344 | 54 | ||
| XM_017358486.2 | XP_017213975.1 | 2 - 3 | 344 | 54 | ||
| GenBank mRNA | AB030900.1 | - | 2 - 3 | 300 | 54 | |
| BC095795.1 | - | 2 - 3 | 281 | 54 | ||
| BC164591.1 | - | 1 - 2 | 155 | 54 | ||
More Information
Set Membership: |
|

mapk8b
Genomic Map