Assay Details
Gene Symbol: mapk8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131721.2 | NP_571796.1 | 6 - 7 | 788 | 62 | |
XM_017358485.2 | XP_017213974.1 | 6 - 7 | 835 | 62 | ||
GenBank mRNA | AB030900.1 | - | 6 - 7 | 791 | 62 | |
BC095795.1 | - | 6 - 7 | 772 | 62 | ||
BC164591.1 | - | 5 - 6 | 646 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
