Assay Details
Gene Symbol: mapk8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131721.2 | NP_571796.1 | 10 - 11 | 1231 | 73 | |
XM_017358485.2 | XP_017213974.1 | 10 - 11 | 1278 | 73 | ||
XM_017358486.2 | XP_017213975.1 | 10 - 11 | 1278 | 73 | ||
GenBank mRNA | AB030900.1 | - | 10 - 11 | 1234 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
