Assay Details
Gene Symbol: aif1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198870.1 | NP_942571.1 | 5 - 6 | 465 | 87 | |
GenBank mRNA | BC059529.1 | - | 5 - 6 | 465 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
