Assay Details
| Gene Symbol: ccnb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199430.1 | NP_955462.1 | 4 - 5 | 631 | 71 | |
| XM_005168860.4 | XP_005168917.1 | 4 - 5 | 708 | 71 | ||
| GenBank mRNA | BC045937.1 | - | 4 - 5 | 631 | 71 | |
| BC066507.1 | - | 4 - 5 | 665 | 71 | ||
| BC164706.1 | - | 4 - 5 | 593 | 71 | ||
More Information
Set Membership: |
|

ccnb2
Genomic Map