Assay Details
| Gene Symbol: elovl6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199532.1 | NP_955826.1 | 1 - 2 | 213 | 77 | |
| GenBank mRNA | BC059459.1 | - | 1 - 2 | 213 | 77 | |
More Information
Set Membership: |
|

elovl6
Genomic Map