Assay Details
| Gene Symbol: ddx39aa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001328342.1 | NP_001315271.1 | 3 - 4 | 413 | 97 | |
| GenBank mRNA | BC042330.1 | - | 4 - 5 | 327 | 97 | |
| BC071505.1 | - | 3 - 4 | 415 | 97 | ||
| BC164154.1 | - | 1 - 2 | 124 | 97 | ||
| EH433199.1 | - | 3 - 4 | 427 | 97 | ||
More Information
Set Membership: |
|

ddx39aa
Genomic Map