Assay Details
| Gene Symbol: rpl27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199724.1 | NP_956018.1 | 3 - 4 | 285 | 154 | |
| GenBank mRNA | BC045965.1 | - | 3 - 4 | 285 | 154 | |
| BC164223.1 | - | 2 - 3 | 286 | 154 | ||
More Information
Set Membership: |
|

rpl27
Genomic Map