Assay Details
| Gene Symbol: psat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199819.2 | NP_956113.1 | 1 - 2 | 189 | 55 | |
| GenBank mRNA | BC044467.1 | - | 1 - 2 | 193 | 55 | |
| EH432914.1 | - | 1 - 2 | 221 | 55 | ||
More Information
Set Membership: |
|

psat1
Genomic Map