Assay Details
| Gene Symbol: brf1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199889.1 | NP_956183.1 | 14 - 15 | 1708 | 116 | |
| GenBank mRNA | BC060523.1 | - | 14 - 15 | 1708 | 116 | |
More Information
Set Membership: |
|

brf1b
Genomic Map