Assay Details
Gene Symbol: brf1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199889.1 | NP_956183.1 | 6 - 7 | 770 | 63 | |
GenBank mRNA | BC060523.1 | - | 6 - 7 | 770 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
