Assay Details
| Gene Symbol: chmp1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200014.1 | NP_956308.1 | 6 - 7 | 610 | 105 | |
| XM_009301289.3 | XP_009299564.1 | 6 - 7 | 647 | 105 | ||
| GenBank mRNA | AY398326.1 | - | 6 - 7 | 616 | 105 | |
| BC045934.1 | - | 6 - 7 | 610 | 105 | ||
| BC065462.1 | - | 6 - 7 | 593 | 105 | ||
| BC067569.1 | - | 6 - 7 | 586 | 105 | ||
| BC153497.1 | - | 6 - 7 | 614 | 105 | ||
| BC165138.1 | - | 6 - 7 | 514 | 105 | ||
More Information
Set Membership: |
|

chmp1b
Genomic Map