Assay Details
| Gene Symbol: mob1ba | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200200.1 | NP_956494.1 | 2 - 3 | 325 | 76 | |
| GenBank mRNA | BC045952.1 | - | 2 - 3 | 325 | 76 | |
| BC063989.1 | - | 2 - 3 | 300 | 76 | ||
| BC165032.1 | - | 1 - 2 | 218 | 76 | ||
More Information
Set Membership: |
|

mob1ba
Genomic Map