Assay Details
Gene Symbol: mob1ba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200200.1 | NP_956494.1 | 3 - 4 | 414 | 128 | |
GenBank mRNA | BC045952.1 | - | 3 - 4 | 414 | 128 | |
BC063989.1 | - | 3 - 4 | 389 | 128 | ||
BC165032.1 | - | 2 - 3 | 307 | 128 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
