Assay Details
| Gene Symbol: tmem170a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200689.1 | NP_956983.1 | 1 - 2 | 143 | 57 | |
| GenBank mRNA | BC059199.1 | - | 1 - 2 | 143 | 57 | |
More Information
Set Membership: |
|

tmem170a
Genomic Map