Assay Details
Gene Symbol: gorasp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200703.2 | NP_956997.1 | 7 - 8 | 945 | 59 | |
XM_021476811.1 | XP_021332486.1 | 7 - 8 | 930 | 59 | ||
GenBank mRNA | AY398354.1 | - | 7 - 8 | 945 | 59 | |
BC059447.1 | - | 7 - 8 | 906 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
