Assay Details
| Gene Symbol: gpr143 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200822.2 | NP_957116.2 | 3 | 601 | 71 | |
| XM_009305016.3 | XP_009303291.1 | 3 | 693 | 71 | ||
| GenBank mRNA | BC059806.1 | - | 3 | 528 | 71 | |
| BC065340.1 | - | 3 | 601 | 71 | ||
More Information
Set Membership: |
|

gpr143
Genomic Map