Assay Details
Gene Symbol: ppm1g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201488.1 | NP_958896.1 | 7 - 8 | 1229 | 80 | |
XM_005169512.4 | XP_005169569.1 | 6 - 7 | 1397 | 80 | ||
GenBank mRNA | BC052132.1 | - | 7 - 8 | 1229 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
