Assay Details
| Gene Symbol: aldh9a1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201508.1 | NP_958916.1 | 11 - 12 | 1503 | 61 | |
| XM_005163231.4 | XP_005163288.1 | 11 - 12 | 1537 | 61 | ||
| XM_009297055.3 | XP_009295330.1 | 11 - 12 | 1548 | 61 | ||
| GenBank mRNA | BC047176.1 | - | 11 - 12 | 1503 | 61 | |
| BC165758.1 | - | 10 - 11 | 1491 | 61 | ||
More Information
Set Membership: |
|

aldh9a1b
Genomic Map