Assay Details
| Gene Symbol: aldh9a1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201508.1 | NP_958916.1 | 7 - 8 | 979 | 62 | |
| XM_005163231.4 | XP_005163288.1 | 7 - 8 | 1013 | 62 | ||
| XM_009297055.3 | XP_009295330.1 | 7 - 8 | 1024 | 62 | ||
| GenBank mRNA | BC047176.1 | - | 7 - 8 | 979 | 62 | |
| BC165758.1 | - | 6 - 7 | 967 | 62 | ||
More Information
Set Membership: |
|

aldh9a1b
Genomic Map