Assay Details
| Gene Symbol: sec61a1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201577.1 | NP_963871.1 | 9 - 10 | 1043 | 80 | |
| GenBank mRNA | AY034614.1 | - | 9 - 10 | 1043 | 80 | |
| BC048881.1 | - | 9 - 10 | 1044 | 80 | ||
| BC165352.1 | - | 8 - 9 | 1009 | 80 | ||
More Information
Set Membership: |
|

sec61a1l
Genomic Map