Assay Details
| Gene Symbol: cacna1da | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203484.1 | NP_982351.1 | 30 - 31 | 3836 | 65 | |
| XM_021479576.1 | XP_021335251.1 | 30 - 31 | 4024 | 65 | ||
| XM_021479577.1 | XP_021335252.1 | 30 - 31 | 4024 | 65 | ||
| XM_021479578.1 | XP_021335253.1 | 30 - 31 | 4024 | 65 | ||
| XM_021479579.1 | XP_021335254.1 | 30 - 31 | 4024 | 65 | ||
| XM_021479580.1 | XP_021335255.1 | 30 - 31 | 4023 | 65 | ||
| XM_021479581.1 | XP_021335256.1 | 29 - 30 | 3948 | 65 | ||
| XM_021479582.1 | XP_021335257.1 | 28 - 29 | 3643 | 65 | ||
| XM_021479583.1 | XP_021335258.1 | 30 - 31 | 4024 | 65 | ||
| XM_021479584.1 | XP_021335259.1 | 29 - 30 | 3948 | 65 | ||
| GenBank mRNA | AY528224.1 | - | 30 - 31 | 3836 | 65 | |
| BC163679.1 | - | 28 - 29 | 3643 | 65 | ||
More Information
Set Membership: |
|

cacna1da
Genomic Map