Assay Details
| Gene Symbol: cacna1da | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203484.1 | NP_982351.1 | 31 - 32 | 3923 | 80 | |
| XM_021479580.1 | XP_021335255.1 | 31 - 32 | 4110 | 80 | ||
| XM_021479584.1 | XP_021335259.1 | 30 - 31 | 4035 | 80 | ||
| GenBank mRNA | AY528224.1 | - | 31 - 32 | 3923 | 80 | |
More Information
Set Membership: |
|

cacna1da
Genomic Map