Assay Details
| Gene Symbol: cacna1da | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203484.1 | NP_982351.1 | 41 - 42 | 5050 | 68 | |
| XM_021479576.1 | XP_021335251.1 | 42 - 43 | 5271 | 68 | ||
| XM_021479577.1 | XP_021335252.1 | 42 - 43 | 5271 | 68 | ||
| XM_021479578.1 | XP_021335253.1 | 42 - 43 | 5271 | 68 | ||
| XM_021479579.1 | XP_021335254.1 | 42 - 43 | 5271 | 68 | ||
| XM_021479580.1 | XP_021335255.1 | 41 - 42 | 5237 | 68 | ||
| XM_021479581.1 | XP_021335256.1 | 41 - 42 | 5195 | 68 | ||
| XM_021479582.1 | XP_021335257.1 | 40 - 41 | 4890 | 68 | ||
| XM_021479584.1 | XP_021335259.1 | 40 - 41 | 5162 | 68 | ||
| GenBank mRNA | AY528224.1 | - | 41 - 42 | 5050 | 68 | |
| BC163679.1 | - | 40 - 41 | 4890 | 68 | ||
More Information
Set Membership: |
|

cacna1da
Genomic Map