Assay Details
| Gene Symbol: psenen | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205576.1 | NP_991139.1 | 3 - 4 | 291 | 64 | |
| GenBank mRNA | AF512427.1 | - | 3 - 4 | 291 | 64 | |
| BC092674.1 | - | 3 - 4 | 249 | 64 | ||
| BC164817.1 | - | 2 - 3 | 190 | 64 | ||
More Information
Set Membership: |
|

psenen
Genomic Map