Assay Details
Gene Symbol: cbx8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205616.2 | NP_991179.2 | 3 - 4 | 228 | 146 | |
GenBank mRNA | AY179330.1 | - | 3 - 4 | 223 | 146 | |
AY398319.1 | - | 3 - 4 | 146 | |||
BC125904.1 | - | 3 - 4 | 228 | 146 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
