Assay Details
| Gene Symbol: amdhd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205681.1 | NP_991244.1 | 10 - 11 | 1124 | 67 | |
| XM_005163743.4 | XP_005163800.1 | 10 - 11 | 1164 | 67 | ||
| XM_005163744.4 | XP_005163801.1 | 10 - 11 | 1127 | 67 | ||
| XM_021469936.1 | XP_021325611.1 | 10 - 11 | 1088 | 67 | ||
| XM_021469942.1 | XP_021325617.1 | 10 - 11 | 1088 | 67 | ||
| GenBank mRNA | BC065449.1 | - | 10 - 11 | 1124 | 67 | |
More Information
Set Membership: |
|

amdhd2
Genomic Map