Assay Details
Gene Symbol: nptnb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205705.1 | NP_991268.1 | 2 - 3 | 721 | 95 | |
GenBank mRNA | BC065957.1 | - | 2 - 3 | 721 | 95 | |
BC155315.1 | - | 2 - 3 | 681 | 95 | ||
CU458926.1 | - | 2 - 3 | 727 | 95 | ||
CU638753.1 | - | 2 - 3 | 719 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
