Assay Details
Gene Symbol: rab22a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205719.2 | NP_991282.2 | 2 - 3 | 355 | 117 | |
GenBank mRNA | BC085393.1 | - | 2 - 3 | 355 | 117 | |
BC165262.1 | - | 2 - 3 | 147 | 117 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
