Assay Details
| Gene Symbol: tuba5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212772.1 | NP_997937.1 | 3 - 4 | 486 | 147 | |
| GenBank mRNA | AY391468.1 | - | 3 - 4 | 481 | 147 | |
| BC067582.1 | - | 3 - 4 | 486 | 147 | ||
More Information
Set Membership: |
|

tuba5
Genomic Map