Assay Details
| Gene Symbol: mdh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213131.1 | NP_998296.1 | 7 - 8 | 812 | 66 | |
| GenBank mRNA | BC053272.1 | - | 7 - 8 | 812 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

mdh2| Gene Symbol: mdh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213131.1 | NP_998296.1 | 7 - 8 | 812 | 66 | |
| GenBank mRNA | BC053272.1 | - | 7 - 8 | 812 | 66 | |
Set Membership: |
|