Assay Details
Gene Symbol: ywhabb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213145.1 | NP_998310.1 | 3 - 4 | 481 | 123 | |
GenBank mRNA | BC053247.1 | - | 3 - 4 | 481 | 123 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
