Assay Details
| Gene Symbol: hprt1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002056.1 | NP_001002056.1 | 6 - 7 | 541 | 118 | |
| GenBank mRNA | BC071336.1 | - | 6 - 7 | 541 | 118 | |
More Information
Set Membership: |
|

hprt1l
Genomic Map