Assay Details
| Gene Symbol: arr3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002405.1 | NP_001002405.1 | 5 - 6 | 242 | 74 | |
| GenBank mRNA | BC076177.1 | - | 5 - 6 | 242 | 74 | |
More Information
Set Membership: |
|

arr3a
Genomic Map