Assay Details
| Gene Symbol: arr3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002405.1 | NP_001002405.1 | 6 - 7 | 427 | 80 | |
| GenBank mRNA | AY900006.1 | - | 1 - 2 | 179 | 80 | |
| BC076177.1 | - | 6 - 7 | 427 | 80 | ||
More Information
Set Membership: |
|

arr3a
Genomic Map