Assay Details
| Gene Symbol: asb13a.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002528.1 | NP_001002528.1 | 4 - 5 | 608 | 68 | |
| XM_009300119.3 | XP_009298394.1 | 3 - 4 | 765 | 68 | ||
| GenBank mRNA | BC076370.1 | - | 4 - 5 | 608 | 68 | |
More Information
Set Membership: |
|

asb13a.1
Genomic Map