Assay Details
Gene Symbol: sult1st6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002599.1 | NP_001002599.1 | 5 - 6 | 948 | 101 | |
GenBank mRNA | BC075996.1 | - | 5 - 6 | 948 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
