Assay Details
| Gene Symbol: srsf5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002610.1 | NP_001002610.1 | 7 - 8 | 606 | 78 | |
| XM_009294706.3 | XP_009292981.1 | 3 - 4 | 596 | 78 | ||
| GenBank mRNA | BC075982.1 | - | 7 - 8 | 606 | 78 | |
More Information
Set Membership: |
|

srsf5b
Genomic Map