Assay Details
Gene Symbol: med19b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003416.2 | NP_001003416.2 | 1 - 2 | 304 | 84 | |
GenBank mRNA | AY648741.1 | - | 1 - 2 | 210 | 84 | |
BC092743.1 | - | 1 - 2 | 304 | 84 | ||
BC165088.1 | - | 1 - 2 | 244 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
