Assay Details
| Gene Symbol: mfsd2ab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003570.2 | NP_001003570.1 | 9 - 10 | 1215 | 88 | |
| GenBank mRNA | BC077117.1 | - | 9 - 10 | 1162 | 88 | |
More Information
Set Membership: |
|

mfsd2ab
Genomic Map