Assay Details
| Gene Symbol: slc35d1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003877.1 | NP_001003877.1 | 7 - 8 | 602 | 125 | |
| GenBank mRNA | AY648822.1 | - | 7 - 8 | 602 | 125 | |
| BC092185.1 | - | 7 - 8 | 576 | 125 | ||
More Information
Set Membership: |
|

slc35d1a
Genomic Map