Assay Details
| Gene Symbol: epb41l3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003987.1 | NP_001003987.1 | 13 - 14 | 1695 | 95 | |
| XM_021470142.1 | XP_021325817.1 | 14 - 15 | 1807 | 95 | ||
| XM_021470143.1 | XP_021325818.1 | 13 - 14 | 1761 | 95 | ||
| XM_021470144.1 | XP_021325819.1 | 14 - 15 | 1806 | 95 | ||
| XM_021470145.1 | XP_021325820.1 | 14 - 15 | 1799 | 95 | ||
| XM_021470146.1 | XP_021325821.1 | 14 - 15 | 1979 | 95 | ||
| XM_021470147.1 | XP_021325822.1 | 14 - 15 | 1838 | 95 | ||
| XM_021470148.1 | XP_021325823.1 | 15 - 16 | 1919 | 95 | ||
| XM_021470149.1 | XP_021325824.1 | 14 - 15 | 1756 | 95 | ||
| XM_021470150.1 | XP_021325825.1 | 14 - 15 | 1804 | 95 | ||
| XM_021470151.1 | XP_021325826.1 | 14 - 15 | 1803 | 95 | ||
| XM_021470152.1 | XP_021325827.1 | 14 - 15 | 1805 | 95 | ||
| XM_021470153.1 | XP_021325828.1 | 13 - 14 | 1754 | 95 | ||
| XM_021470154.1 | XP_021325829.1 | 13 - 14 | 1701 | 95 | ||
| XM_021470155.1 | XP_021325830.1 | 3 - 4 | 426 | 95 | ||
| GenBank mRNA | BC080246.1 | - | 13 - 14 | 1695 | 95 | |
| BC158197.1 | - | 13 - 14 | 1701 | 95 | ||
More Information
Set Membership: |
|

epb41l3a
Genomic Map