Assay Details
| Gene Symbol: epb41l3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003987.1 | NP_001003987.1 | 17 - 18 | 2176 | 66 | |
| XM_021470142.1 | XP_021325817.1 | 19 - 20 | 2843 | 66 | ||
| XM_021470143.1 | XP_021325818.1 | 18 - 19 | 2797 | 66 | ||
| XM_021470144.1 | XP_021325819.1 | 19 - 20 | 2842 | 66 | ||
| XM_021470145.1 | XP_021325820.1 | 19 - 20 | 2835 | 66 | ||
| XM_021470146.1 | XP_021325821.1 | 19 - 20 | 3015 | 66 | ||
| XM_021470147.1 | XP_021325822.1 | 19 - 20 | 2874 | 66 | ||
| XM_021470148.1 | XP_021325823.1 | 20 - 21 | 2955 | 66 | ||
| XM_021470149.1 | XP_021325824.1 | 19 - 20 | 2792 | 66 | ||
| XM_021470150.1 | XP_021325825.1 | 19 - 20 | 2840 | 66 | ||
| XM_021470151.1 | XP_021325826.1 | 19 - 20 | 2839 | 66 | ||
| XM_021470152.1 | XP_021325827.1 | 18 - 19 | 2286 | 66 | ||
| XM_021470153.1 | XP_021325828.1 | 17 - 18 | 2235 | 66 | ||
| XM_021470154.1 | XP_021325829.1 | 17 - 18 | 2182 | 66 | ||
| XM_021470155.1 | XP_021325830.1 | 8 - 9 | 1462 | 66 | ||
| GenBank mRNA | BC080246.1 | - | 17 - 18 | 2176 | 66 | |
| BC158197.1 | - | 17 - 18 | 2182 | 66 | ||
More Information
Set Membership: |
|

epb41l3a
Genomic Map