Assay Details
| Gene Symbol: clpxa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004581.1 | NP_001004581.1 | 10 - 11 | 1777 | 72 | |
| XM_021477471.1 | XP_021333146.1 | 11 - 12 | 1887 | 72 | ||
| GenBank mRNA | BC081643.1 | - | 10 - 11 | 1777 | 72 | |
More Information
Set Membership: |
|

clpxa
Genomic Map